missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KRT23 Monoclonal antibody specifically detects KRT23 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | KRT23 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2F6 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CK23, CK-23, cytokeratin 23, Cytokeratin-23, DKFZp434G032, HAIK1, hyperacetylation-inducible type I keratin, K23histone deacetylase inducible keratin 23, keratin 23 (histone deacetylase inducible), keratin, type I cytoskeletal 23, keratin-23, MGC26158, type I intermediate filament cytokeratin |
| Host Species | Mouse |
| Immunogen | Recombinant human KRT23 (271-422aa) purified from E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?