missing translation for 'onlineSavingsMsg'
Learn More

KRT23 Antibody (2F6), DyLight 755, Novus Biologicals™

Product Code. 18402416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402416 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402416 Supplier Novus Biologicals Supplier No. NBP222590IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KRT23 Monoclonal antibody specifically detects KRT23 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT23
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2F6
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CK23, CK-23, cytokeratin 23, Cytokeratin-23, DKFZp434G032, HAIK1, hyperacetylation-inducible type I keratin, K23histone deacetylase inducible keratin 23, keratin 23 (histone deacetylase inducible), keratin, type I cytoskeletal 23, keratin-23, MGC26158, type I intermediate filament cytokeratin
Host Species Mouse
Immunogen Recombinant human KRT23 (271-422aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25984
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.