missing translation for 'onlineSavingsMsg'
Learn More

KRR1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18409226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409226 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409226 Supplier Novus Biologicals Supplier No. NBP222252DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRR1 Polyclonal antibody specifically detects KRR1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRR1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HIV-1 Rev binding protein 2, HIV-1 Rev-binding protein 2, HRB2, KRR1 small subunit processome component homolog, KRR1, small subunit (SSU) processome component, homolog (yeast), Rev interacting protein, Rev-interacting protein 1, Rip-1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 300 and 350 of human Small Subunit (SSU) Processome Component using the numbering given in entry NP_008974.5 (GeneID 11103).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11103
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.