Learn More
Spectrum Labs™ KrosFlo KR1 Auxiliary Pump for Diafiltration
Product Code. 15777586
Change view
Click to view available options
For Use With (Equipment):
KR2i/KMPi TFF Systems
Product Code. For Use With (Equipment)
15777586 KR2i/KMPi TFF Systems
1 options
Product Code. 15777586 Supplier Spectrum Labs™ Supplier No. ACR1U2001N
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Spectrum Labs™ KrosFlo KR1 Auxiliary Pump for Diafiltration is specifically designed to facilitate the KR2i/KMPi TFF system diafiltration process.
  • Auxiliary pump for diafiltration
  • Suitable for KR2i/KMPi TFF systems
Specifications
For Use With (Equipment) KR2i/KMPi TFF Systems
Product Type Diafiltration Pump, 600 rpm