missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRIT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | KRIT1 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18332186
|
Novus Biologicals
NBP3-05054-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18393385
|
Novus Biologicals
NBP3-05054-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KRIT1 Polyclonal specifically detects KRIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| KRIT1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 889 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 467-736 of human KRIT1 (NP_919438.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title