missing translation for 'onlineSavingsMsg'
Learn More

KRIT1 Antibody (1D8), Novus Biologicals™

Product Code. 18329717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329717 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329717 Supplier Novus Biologicals Supplier No. H00000889M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KRIT1 Monoclonal antibody specifically detects KRIT1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRIT1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1D8
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004903
Gene Alias CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing
Host Species Mouse
Immunogen KRIT1 (NP_004903, 637 a.a. ~ 736 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AAFFTGQIFTKASPSNHKVIPVYVGVNIKGLHLLNMETKALLISLKYGCFMWQLGDTDTCFQIHSMENKMSFIVHTKQAGLVVKLLMKLNGQLMPTERNS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 889
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.