missing translation for 'onlineSavingsMsg'
Learn More

Kresoxim-methyl-d7, TRC

Product Code. 30399022
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399022 1 mg 1mg
30393487 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399022 Supplier Toronto Research Chemicals Supplier No. TRCK6590021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kresoxim-methyl-d7
Purity Grade Notes HPLC
Molecular Formula C18H12D7NO4
Quantity 1 mg
Synonym (αE)-α-(methoxyimino)-2-[(2-methylphenoxy)methyl]benzeneacetic Acid Methyl Ester-d7, BAS 490-02F-d7, BAS 490F-d7, Candit-d7, Cygnus-d7, Discus-d7, Discus (fungicide)-d7, RIL-FA 200-d7, Strob 1-d7, Stroby-d7, Stroby WG-d7
InChI Formula InChI=1S/C18H19NO4/c1-13-8-4-7-11-16(13)23-12-14-9-5-6-10-15(14)17(19-22-3)18(20)21-2/h4-11H,12H2,1-3H3/b19-17-/i1D3,4D,7D,8D,11D
SMILES [2H]C1=C([2H])C([2H])=C([2H])C(C([2H])([2H])[2H])=C1OCC2=C(/C(C(OC)=O)=N\OC)C=CC=C2
Molecular Weight (g/mol) 320.39
Percent Purity >95
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.