missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kresoxim-methyl-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Kresoxim-methyl-d7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H12D7NO4 |
| Quantity | 10 mg |
| Synonym | (αE)-α-(methoxyimino)-2-[(2-methylphenoxy)methyl]benzeneacetic Acid Methyl Ester-d7, BAS 490-02F-d7, BAS 490F-d7, Candit-d7, Cygnus-d7, Discus-d7, Discus (fungicide)-d7, RIL-FA 200-d7, Strob 1-d7, Stroby-d7, Stroby WG-d7 |
| InChI Formula | InChI=1S/C18H19NO4/c1-13-8-4-7-11-16(13)23-12-14-9-5-6-10-15(14)17(19-22-3)18(20)21-2/h4-11H,12H2,1-3H3/b19-17-/i1D3,4D,7D,8D,11D |
| SMILES | [2H]C1=C([2H])C([2H])=C([2H])C(C([2H])([2H])[2H])=C1OCC2=C(/C(C(OC)=O)=N\OC)C=CC=C2 |
| Molecular Weight (g/mol) | 320.39 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?