missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KREMEN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534520
This item is not returnable.
View return policy
Description
A suggested positive control is rat small intestine tissue lysate. PA5-34520 can be used with blocking peptide PEP-1563.
KREMEN1 is a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. This protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains.
Specifications
| KREMEN1 | |
| Polyclonal | |
| Unconjugated | |
| Kremen1 | |
| AV002070; dickkopf receptor; KREMEN; kremen protein 1; KREMEN1; kringle containing transmembrane protein 1; kringle domain-containing transmembrane protein 1; kringle-coding gene marking the eye and the nose; kringle-containing protein marking the eye and the nose; KRM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114107, 83999, 84035 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q924S4, Q96MU8, Q99N43 | |
| Kremen1 | |
| An 18 amino acid peptide near the carboxy terminus of human KREMEN1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction