missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kremen-2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22339PCP
This item is not returnable.
View return policy
Description
Kremen-2 Polyclonal antibody specifically detects Kremen-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Kremen-2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| Dickkopf receptor 2, kremen protein 2, kringle containing transmembrane protein 2, Kringle domain-containing transmembrane protein 2, KRM2Kringle-containing protein marking the eye and the nose, MGC10791, MGC16709 | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human Kremen-2. The immunogen is located within amino acids 390-440 of Kremen-2. Amino Acid Sequence: ALGASRGGPRRSWAV | |
| 0.1 mL | |
| Signal Transduction | |
| 79412 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction