missing translation for 'onlineSavingsMsg'
Learn More

Kremen-2 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30655306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30655306 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30655306 Supplier Novus Biologicals Supplier No. AF1764JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Kremen-2 Polyclonal antibody specifically detects Kremen-2 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kremen-2
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Dickkopf receptor 2, kremen protein 2, kringle containing transmembrane protein 2, Kringle domain-containing transmembrane protein 2, KRM2Kringle-containing protein marking the eye and the nose, MGC10791, MGC16709
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kremen-2, Gly25-Thr371, Accession # Q8K1S7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79412
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.