missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kremen-1 Antibody (RM0120-6K45), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22498F
This item is not returnable.
View return policy
Description
Kremen-1 Monoclonal antibody specifically detects Kremen-1 in Mouse samples. It is validated for Western Blot
Specifications
| Kremen-1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0120-6K45 | |
| Western Blot | |
| EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1Dickkopf receptor | |
| Mouse recombinant protein of the extracellular domain of Kremen-1 | |
| 0.1 mL | |
| Signal Transduction | |
| 83999 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction