missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRBA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565620
This item is not returnable.
View return policy
Description
Immunogen sequence: DLQGLSRGTAR RARPLPPDAP PAEPPGLHCS SSQQLLSSTP SCHAAPPAHP LLAHTGGHQS PLPPLVPAAL PLQGASPPAA SADADVPTSG VAPDGIPERP KEPSSLLGGV QRALQEELWG GEHRDPRW Highest antigen sequence identity to the following orthologs - mouse 50%, rat 48%.
KRBA1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
| KRBA1 | |
| Polyclonal | |
| Unconjugated | |
| KRBA1 | |
| KIAA1862; KRAB-A domain containing 1; KRBA1; Protein KRBA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A5PL33 | |
| KRBA1 | |
| Recombinant Human KRBA1. Recombinant protein control fragment (Product #RP-104903). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction