missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRAS Antibody (OTI2C1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71079CL1
This item is not returnable.
View return policy
Description
KRAS Monoclonal antibody specifically detects KRAS in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Western Blot
Specifications
| KRAS | |
| Monoclonal | |
| CoraFluor 1 | |
| cellular c-Ki-ras2 proto-oncogene, c-Ki-ras, C-K-RAS, GTPase KRas, Ki-Ras, Kirsten rat sarcoma-2 viral (v-Ki-ras2) oncogene homolog, K-Ras 2, K-ras p21 protein, KRAS1, K-RAS2A, K-RAS2B, KRAS2NS3, K-RAS4A, K-RAS4B, NS, oncogene KRAS2, PR310 c-K-ras oncogene, RASK2c-Kirsten-ras protein, transforming protein p21, v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog, v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog | |
| Full length human recombinant protein of human KRAS (NP_203524) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 3845 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot | |
| OTI2C1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction