missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRAB related Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116309
This item is not returnable.
View return policy
Description
KRAB related Polyclonal Antibody for Western Blot
SSX11 could act as a modulator of transcription.
Specifications
| KRAB related | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of KRAB-related domain-containing protein. | |
| 400 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction