missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KPNA6 Polyclonal specifically detects KPNA6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | KPNA6 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | importin alpha 7 subunit, importin subunit alpha-7, importin-alpha-S2, IPOA7FLJ11249, karyopherin alpha 6 (importin alpha 7), Karyopherin subunit alpha-6, KPNA7, MGC17918 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6 (NP_036448.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?