missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KPNA6 Polyclonal Antibody

Description
A suggested positive control is 293 cell lysate. PA5-21036 can be used with blocking peptide PEP-1150.
Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA6 belongs to a subfamily within the KPNA family that also includes IPOA5&6. Down-regulation of KPNA6 by RNAi expression in HeLa cells strongly inhibited cell proliferation, possibly due to blocking the nuclear import of specific factors essential for cell growth and proliferation. KPNA6 also interacts with the Rev protein of HIV-1 and promote its nuclear import.
Specifications
Specifications
| Antigen | KPNA6 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KPNA6 |
| Gene Accession No. | O35345, O60684 |
| Gene Alias | FLJ11249; importin alpha 7 subunit; importin alpha-S2; Importin subunit alpha-7; importin-alpha-S2; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 6; karyopherin alpha 6 (importin alpha 7); karyopherin subunit alpha 6; karyopherin subunit alpha-6; Kpna5; Kpna6; KPNA7; MGC17918; NPI-2; RP4-622L5.1 |
| Gene Symbols | KPNA6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?