missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA5 Polyclonal Antibody

KPNA5 Polyclonal Antibody
Brand: Invitrogen PA572750
This item is not returnable.
View return policy
Description
KPNA5 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| KPNA5 | |
| Polyclonal | |
| Unconjugated | |
| Kpna5 | |
| hypothetical protein LOC553205; im:7151662; importin alpha 6; importin alpha-S2; Importin subunit alpha-6; Importin subunit alpha-6-like protein; importin subunit alpha-7; Ipoa6; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 5; karyopherin alpha 5 (importin alpha 6); karyopherin subunit alpha 5; karyopherin subunit alpha-5; Karyopherin subunit alpha-6; kpna5; Kpna6; NPI-2; SRP6; zgc:110662 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O15131 | |
| Kpna5 | |
| KPNA5 antibody for an 18 amino acid synthetic peptide near the amino terminus of human KPNA5. The immunogen is within amino acids 50 - 100 of KPNA5. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction