missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total KPNA5.
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC) which consists of 60-100 proteins and is probably 120 million daltons in molecular size. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion; larger molecules are transported by an active process. Most nuclear proteins contain short basic amino acid sequences known as nuclear localization signals (NLSs). KPNA5 protein belongs to the importin alpha protein family and is thought to be involved in NLS-dependent protein import into the nucleus.
Specifications
Specifications
| Antigen | KPNA5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | Kpna5 |
| Gene Accession No. | O15131, Q56R16 |
| Gene Alias | hypothetical protein LOC553205; im:7151662; importin alpha 6; importin alpha-S2; Importin subunit alpha-6; Importin subunit alpha-6-like protein; importin subunit alpha-7; Ipoa6; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 5; karyopherin alpha 5 (importin alpha 6); karyopherin subunit alpha 5; karyopherin subunit alpha-5; Karyopherin subunit alpha-6; kpna5; Kpna6; NPI-2; SRP6; zgc:110662 |
| Gene Symbols | Kpna5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
