missing translation for 'onlineSavingsMsg'
Learn More

KPNA5 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18424435 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424435 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424435 Supplier Novus Biologicals Supplier No. NBP281835AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KPNA5 Polyclonal antibody specifically detects KPNA5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA5
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias importin alpha 6, importin subunit alpha-6, IPOA6, karyopherin alpha 5 (importin alpha 6), Karyopherin subunit alpha-5, SRP6
Host Species Rabbit
Immunogen KPNA5 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human KPNA5. The immunogen is located within amino acids 50 - 100 of KPNA5. Amino Acid Squence: LESPIQDPDISSTVPIPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3841
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.