Learn More
KPNA3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28050-1-AP-150UL
Description
The KPNA3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KPNA3. This antibody recognizes human, mouse antigen. The KPNA3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| KPNA3 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| hSRP1, Importin alpha Q2, Importin subunit alpha 3, IPOA4, Karyopherin subunit alpha 3, KPNA3, Qip2, SRP1, SRP1 gamma, SRP1gamma, SRP4 | |
| 150 μL | |
| 3839 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |