missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KPNA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12809182
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809182 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809182 Supplier Invitrogen™ Supplier No. PA521032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-21032 can be used with blocking peptide PEP-1146.

Recombination activating proteins RAG1 and RAG2 regulate and mediate VJ recombination, the process by which genes for immunoglobulins and T-cell receptors are generated. Several other ubiquitously expressed proteins are thought to be recruited in the recombination process. Among these are the genes affected in severe combined immune deficiency and genes involved in ds-DNA break repair. The protein encoded by this gene interacts with RAG1 and may play a role in VJ recombination. Two transcript variants, one protein-coding and the other not, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KPNA1
Gene Accession No. P52294, P83953, Q60960
Gene Alias AW494490; IMA2; Importin alpha 2; importin alpha 5; Importin alpha P1; importin alpha-5; importin alpha-S1; importin subunit alpha-1; importin subunit alpha-5; Importin subunit alpha-5, N-terminally processed; importin-alpha-S1; IPOA 1; IPOA1; IPOA5; karyopherin (importin) alpha 1; Karyopherin A2; karyopherin alpha 1; karyopherin alpha 1 (importin alpha 5); karyopherin subunit alpha 1; karyopherin subunit alpha-1; karyopherin/importin alpha-1; KPNA1; KPNA2; m-importin-alpha-S1; mSRP1; NPI1; NPI-1; Nucleoprotein interactor 1; RAG cohort 1; RAG cohort protein 2; RCH 1; RCH1; RCH2; recombination activating gene cohort 2; SRP 1; SRP1; SRP1 alpha; SRP1-beta
Gene Symbols KPNA1
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of human KPNA1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16646, 288064, 3836
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.