missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KPNA1 Monoclonal Antibody (K94035_3A10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556064
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: LKSYKNKSLN PDEMRRRREE EGLQLRKQKR EEQLFKRRNV ATAEEETEEE VMSDGGFHEA QISNMEMAPG GVITSDMIEM
Recombination activating proteins RAG1 and RAG2 regulate and mediate VJ recombination, the process by which genes for immunoglobulins and T-cell receptors are generated. Several other ubiquitously expressed proteins are thought to be recruited in the recombination process. Among these are the genes affected in severe combined immune deficiency and genes involved in ds-DNA break repair. The protein encoded by this gene interacts with RAG1 and may play a role in VJ recombination. Two transcript variants, one protein-coding and the other not, have been found for this gene.
Specifications
| KPNA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P52294, Q60960 | |
| KPNA1 | |
| Recombinant human KPNA1 (immunogen range is 11-90). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K94035_3A10 | |
| Unconjugated | |
| KPNA1 | |
| AW494490; IMA2; Importin alpha 2; importin alpha 5; Importin alpha P1; importin alpha-5; importin alpha-S1; importin subunit alpha-1; importin subunit alpha-5; Importin subunit alpha-5, N-terminally processed; importin-alpha-S1; IPOA 1; IPOA1; IPOA5; karyopherin (importin) alpha 1; Karyopherin A2; karyopherin alpha 1; karyopherin alpha 1 (importin alpha 5); karyopherin subunit alpha 1; karyopherin subunit alpha-1; karyopherin/importin alpha-1; KPNA1; KPNA2; m-importin-alpha-S1; mSRP1; NPI1; NPI-1; Nucleoprotein interactor 1; RAG cohort 1; RAG cohort protein 2; RCH 1; RCH1; RCH2; recombination activating gene cohort 2; SRP 1; SRP1; SRP1 alpha; SRP1-beta | |
| Mouse | |
| Protein A | |
| RUO | |
| 16646, 3836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction