missing translation for 'onlineSavingsMsg'
Learn More

KP496, MedChemExpress

Product Code. 16422488
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16422488

Brand: Medchem Express HYU00253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KP496 is a selective, dual antagonist for Leukotriene D4 receptor and Thromboxane A2 receptor.

KP496 is a selective, dual antagonist for Leukotriene D4 receptor and Thromboxane A2 receptor.

Specifications

Chemical Name or Material KP496
CAS 217799-03-6
Color White
Purity Grade Notes Research
Molecular Formula C31H34ClN3O7S3
Quantity 5 mg
SMILES O=C(O)C1=CC=CC=C1S(=O)(N(CCCCNS(=O)(C2=CC=C(Cl)C=C2)=O)CC3=CC=CC(OCC4=NC(C(C)C)=CS4)=C3)=O
Molecular Weight (g/mol) 692.27
Formula Weight 692.27
Percent Purity 95.81%
Grade Research
Physical Form Crystals
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.