missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KOR Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-68860
This item is not returnable.
View return policy
Description
KOR Polyclonal specifically detects KOR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| KOR | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
| Goat | |
| 43 kDa | |
| 0.1 mg | |
| GPCR, Signal Transduction | |
| 4986 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:16000 | |
| P41145 | |
| OPRK1 | |
| Peptide with sequence C-PAYLRDIDGMNKP corresponding to C-Terminus according to NP_000903.2. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction