missing translation for 'onlineSavingsMsg'
Learn More

KOR Antibody (387301) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30172756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30172756 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30172756 Supplier Novus Biologicals Supplier No. FAB3895JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KOR Monoclonal antibody specifically detects KOR in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen KOR
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 387301
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human KOR, Met1-Val380, Accession # P41145
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4986
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.