missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Protects the environment from droplets emitted by the wearer while providing protection from splashes of biological fluids
- Effective against bacteria, particles and viruses (BFE, PFE, and VFE)
- Feature exclusive Origami folding and Smile welding for greater comfort and an ideal fit on the wearer's face
- Softex technology limits irritation to the face and does not wear out with prolonged use, even in contact with beard
- Clinically tested and recommended by AFPRAL (French Association for Allergy Prevention)
- Reduces the onset of symptoms related to seasonal allergies such as asthma, rhinitis, etc. and improves quality of life of pollen allergy sufferers (as found in Alyatec 2022 pilot clinical study)
- Hypoallergenic
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| Color | Green |
| Fastening Type | Earloops |
| Material | Polypropylene |
| Size | One Size |
| Sterility | Non-sterile |
| Style | Pouch |
| Certifications/Compliance | ISO 11737:2018 + A1:2021, ISO 10993-10, ISO 10993-5, ISO 13485, ISO 9001 |
| Certifications/Compliance (Europe) | EU Regulation 2017/745 |
| Disposable | Single-use |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
