missing translation for 'onlineSavingsMsg'
Learn More

Kojic acid, MedChemExpress

Product Code. 16459406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459406 Supplier Medchem Express Supplier No. HYW050154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kojic acid is a natural substance produced by Aspergillus oryzae, also used as an anti-oxidant and radio-protective agent.

Kojic acid is a natural substance produced by Aspergillus oryzae, also used as an anti-oxidant and radio-protective agent.

Specifications

Chemical Name or Material Kojic acid
CAS 501-30-4
Color White
Purity Grade Notes Research
Molecular Formula C6H6O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (703.68 mM) ∣H2O : 50 mg/mL (351.84 mM; Need ultrasonic)
SMILES O=C1C=C(CO)OC=C1O
Molecular Weight (g/mol) 142.11
Formula Weight 142.11
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.