Learn More
Koflo Tube Mixer
Use Tube Mixer to mix fluids as you pump them through a pipe line.
Brand:  Koflo 04669-54
 View more versions of this product
Product Code. 11764119
  • £173.00 / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Able to mix light viscosity to high viscosity components.
  • Accept standard tube fittings.
  • These mixers consist of a series of fixed right- and left-hand elements.
  • Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies.
  • Mixers have plain ends and accept standard tube fittings.
Specifications
Specifications
0.2 in.
7.5 in.
Stainless Steel (316)
Mixing high-viscosity fluids
5 mm
190 mm
Tube Mixer