missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kobe2602, MedChemExpress
Description
Kobe2602 is a Ras-Raf interaction inhibitor. Kobe2602 inhibits the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki of 149 μM. Kobe2602 has antitumor activity.
Specifications
Specifications
| Chemical Name or Material | Kobe2602 |
| CAS | 454453-49-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C14H9F4N5O4S |
| Quantity | 250 mg |
| Solubility Information | DMSO : 14.3 mg/mL (34.10 mM; Need ultrasonic and warming) |
| SMILES | FC(C1=CC([N+]([O-])=O)=C(NNC(NC2=CC=C(F)C=C2)=S)C([N+]([O-])=O)=C1)(F)F |
| Molecular Weight (g/mol) | 419.31 |
| Formula Weight | 419.31 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?