missing translation for 'onlineSavingsMsg'
Learn More

Ko 143, MedChemExpress

Product Code. 16489466
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489466 50 mg 50mg
16422778 10 mg 10mg
16415517 5 mg 5mg
16494226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489466 Supplier Medchem Express Supplier No. HY10010
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. Ko 143 displays >200-fold selectivity over P-gp and MRP-1 transporters.

Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. Ko 143 displays >200-fold selectivity over P-gp and MRP-1 transporters.

Specifications

Chemical Name or Material Ko 143
CAS 461054-93-3
Color White
Purity Grade Notes Research
Molecular Formula C26H35N3O5
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (212.96 mM; Need ultrasonic)
SMILES O=C1N2[C@]([H])(C3=C(C[C@]2(C(N[C@H]1CCC(OC(C)(C)C)=O)=O)[H])C4=CC=C(OC)C=C4N3)CC(C)C
Molecular Weight (g/mol) 469.57
Formula Weight 469.57
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.