missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KNP-I Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527430
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a potential mitochondrial protein that is a member of the DJ-1/PfpI gene family. This protein is overexpressed in fetal Down syndrome brain. Alternate splicing results in multiple transcript variants.
Specifications
| KNP-I | |
| Polyclonal | |
| Unconjugated | |
| GATD3A | |
| C21orf33; chromosome 21 open reading frame 33; ES1; ES1 protein homolog, mitochondrial; GATD3A; GATD3B; Glutamine amidotransferase-like class 1 domain-containing protein 3A, mitochondrial; Glutamine amidotransferase-like class 1 domain-containing protein 3B, mitochondrial; GT335; H KNPH; HES1; human HES1 protein, homolog to E.coli and zebrafish ES1 protein; Keio novel protein I; Keio novel protein-I; KNPH; KNPI; KNP-I; Protein GT335; Protein HES1; testis secretory sperm-binding protein Li 237E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P0DPI2 | |
| GATD3A | |
| Recombinant protein encompassing a sequence within the center region of human C21orf33. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction