missing translation for 'onlineSavingsMsg'
Learn More

KN-62

Product Code. p-7042411
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464819 1 mg 1mg
15474819 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464819 Supplier Thermo Scientific Alfa Aesar Supplier No. J62444.M^
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potently and specifically inhibits calcium/calmodulin kinase II | CAS: 127191-97-3 | C38H35N5O6S2 | 721.85 g/mol

It is used as a potent and selective inhibitor of CaMKII. KN-62 has demonstrated potent and selective inhibition of Ca2+/calmodulin-dependent protein kinase II (CaMKII; Ki=0.9 ?M) which causes cells to arrest in the S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 ?M of KN-62 blocked CaMKII phosphorylation of III-AC, and inhibited hormone-stimulated Ca2+ oscillations. Additionally, KN-62 is a potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a potent and selective inhibitor of CaMKII. KN-62 has demonstrated potent and selective inhibition of Ca2+/calmodulin-dependent protein kinase II (CaMKII; Ki=0.9 μM) which causes cells to arrest in the S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 μM of KN-62 blocked CaMKII phosphorylation of III-AC, and inhibited hormone-stimulated Ca2+ oscillations. Additionally, KN-62 is a potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).

Solubility
Soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (0.93 mg/ml), methanol (10 mg/ml), DMSO (12 mg/ml), chloroform (20 mg/ml), ethanol (∼1 mg/ml), 1:2 DMF:PBS (pH 7.2) (∼0.3 mg/ml), and DMF & DMSO (∼30 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127191-97-3
Molecular Formula C38H35N5O6S2
Molecular Weight (g/mol) 721.85
MDL Number MFCD00083180
InChI Key RJVLFQBBRSMWHX-UHFFFAOYNA-N
Synonym s-4-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, unii-63hm46xpow, chembl28324, 63hm46xpow, 1-n,o-bis 5-isoquinolinesulfonyl-n-methyl-l-tyrosyl-4-phenylpiperazine, s-5-isoquinolinesulfonic acid 4-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl isoquinolinesulfonic acid ester, 4-2s-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, 5-isoquinolinesulfonic acid, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester
PubChem CID 5312126
IUPAC Name [4-[(2S)-2-[isoquinolin-5-ylsulfonyl(methyl)amino]-3-oxo-3-(4-phenylpiperazin-1-yl)propyl]phenyl] isoquinoline-5-sulfonate
SMILES CN(C(CC1=CC=C(OS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1)C(=O)N1CCN(CC1)C1=CC=CC=C1)S(=O)(=O)C1=C2C=CN=CC2=CC=C1

Specifications

Melting Point 65°C to 67°C
Color Yellow
Linear Formula NH2(CH2)10NH2
Quantity 1 mg
Solubility Information Soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (0.93mg/ml),methanol (10mg/ml),DMSO (12mg/ml),chloroform (20mg/ml),ethanol (∽1mg/ml),1:2 DMF:PBS (pH 7.2) (∽0.3mg/ml),and DMF & DMSO (∽30mg/ml).
Formula Weight 721.84
Physical Form Powder
Chemical Name or Material KN-62

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.