missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT3B/NSD1 Antibody (3E6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064324-M06
This item is not returnable.
View return policy
Description
KMT3B/NSD1 Monoclonal antibody specifically detects KMT3B/NSD1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| KMT3B/NSD1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Androgen receptor coactivator 267 kDa protein, Androgen receptor-associated protein of 267 kDa, ARA267STO, DKFZp666C163, EC 2.1.1.43, FLJ10684, FLJ22263, FLJ44628, H3 lysine-36 and H4 lysine-20 specific, KMT3BSotos syndrome, Lysine N-methyltransferase 3B, NR-binding SET domain-containing protein, nuclear receptor binding SET domain protein 1 | |
| NSD1 (NP_071900, 2 a.a. ∽ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DQTCELPRRNCLLPFSNPVNLDAPEDKDSPFGNGQSNFSEPLNGCTMQLSTVSGTSQNAYGQDSPSCYIPLRRLQDLASMINVEYLNGSADGSESFQDPEKSDSRAQT | |
| 0.1 mg | |
| Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
| 64324 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 3E6 | |
| Western Blot 1:500, ELISA | |
| NP_071900 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction