missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KMT2D Recombinant Rabbit Monoclonal Antibody (9H13L17)

Product Code. 15632025
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15632025 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15632025 Supplier Invitrogen™ Supplier No. 701869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Cat, Mouse and Sheep. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

KMT2D is a histone methyltransferase that methylates Lys-4 of histone H3 (H3K4me). H3K4me represents a specific tag for epigenetic transcriptional activation. KMT2D acts as a coactivator for estrogen receptors by being recruited by ESR1 and activating transcription. KMT2D is involved in chromatin organization, chormatin silencig, oocyte growth, oogenesis, and a positive regulation of cell proliferation. Mutations in the KMT2D gene cause Kabuki syndrome (KABUK1) which results in congential mental retardation syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2D
Applications Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9H13L17
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene KMT2D
Gene Accession No. O14686
Gene Alias AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21
Gene Symbols KMT2D
Host Species Rabbit
Immunogen Peptides corresponding to Human KMT2D (aa 2-15, 1071-1082).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.