missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18671-100UL
This item is not returnable.
View return policy
Description
KMT2D Polyclonal antibody specifically detects KMT2D in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| KMT2D | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:1000-1:5000 | |
| O14686 | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| 8085 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Whole antiserum | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| Polyclonal antibody raised in rabbit against mouse KMT2D (Myeloid/lymphoid or mixed-lineage leukemia protein 4), using three KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein (1). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction