missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KMT2D Monoclonal Antibody (3C8E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548639
This item is not returnable.
View return policy
Description
KMT2D Monoclonal Antibody for Western Blot, Flow, ELISA
KMT2D is a histone methyltransferase that methylates Lys-4 of histone H3 (H3K4me). H3K4me represents a specific tag for epigenetic transcriptional activation. KMT2D acts as a coactivator for estrogen receptors by being recruited by ESR1 and activating transcription. KMT2D is involved in chromatin organization, chormatin silencig, oocyte growth, oogenesis, and a positive regulation of cell proliferation. Mutations in the KMT2D gene cause Kabuki syndrome (KABUK1) which results in congential mental retardation syndrome.
Specifications
| KMT2D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O14686 | |
| KMT2D | |
| Purified recombinant fragment of human KMT2D (AA: 445-599) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3C8E11 | |
| Unconjugated | |
| KMT2D | |
| AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8085 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction