missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Antibody (CL5346), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £443.00
Specifications
| Antigen | KMT2D |
|---|---|
| Clone | CL5346 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218112
|
Novus Biologicals
NBP2-61416 |
100 μL |
£443.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696358
|
Novus Biologicals
NBP2-61416-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KMT2D Monoclonal specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| KMT2D | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| MLL2 | |
| IgG1 | |
| Protein A purified |
| CL5346 | |
| Monoclonal | |
| Purified | |
| Human | |
| 8085 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AQLADTLFSKGLGPWDPPDNLAETQKPEQSSLVPGHLDQVNGQVVPEASQLSIKQEPREEPCALGAQSVKREANGEPIGAPGTSNHLLLAGPRSEAGHLLLQKLLRAKNVQLSTGRGSEGLRAEINGHI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title