missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-18667-25UL
This item is not returnable.
View return policy
Description
KMT2B Polyclonal antibody specifically detects KMT2B in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| KMT2B | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:200-1:2000 | |
| Q9UMN6 | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Primary | |
| Mouse | |
| Antisera |
| Western Blot, ELISA | |
| Unconjugated | |
| Whole antiserum | |
| EC 2.1.1.43, histone-lysine N-methyltransferase MLL4, HRX2, KIAA0304WW domain-binding protein 7, Lysine N-methyltransferase 2D, mixed lineage leukemia gene homolog 2, MLL2, myeloid/lymphoid or mixed-lineage leukemia 4, Myeloid/lymphoid or mixed-lineage leukemia protein 4, Trithorax homolog 2, trithorax homologue 2, TRX2, WBP7, WBP-7, WW domain binding protein 7 | |
| Polyclonal antibody raised in rabbit against mouse KMT2B (Myeloid/lymphoid or mixed-lineage leukemia protein 2), using two KLH-conjugated synthetic peptides containing an amino acid sequence from the N-terminal and one peptide containing a sequence from the central part of the protein, respectively. | |
| 25 μg | |
| Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 9757 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction