missing translation for 'onlineSavingsMsg'
Learn More

KMT2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18350883
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350883 100 μg 100µL
18345342 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18350883 Supplier Bio-Techne Supplier No. NBP315609100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KMT2B Polyclonal antibody specifically detects KMT2B in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias EC 2.1.1.43, histone-lysine N-methyltransferase MLL4, HRX2, KIAA0304WW domain-binding protein 7, Lysine N-methyltransferase 2D, mixed lineage leukemia gene homolog 2, MLL2, myeloid/lymphoid or mixed-lineage leukemia 4, Myeloid/lymphoid or mixed-lineage leukemia protein 4, Trithorax homolog 2, trithorax homologue 2, TRX2, WBP7, WBP-7, WW domain binding protein 7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1370-1570 of human KMT2B (NP_055542.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9757
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.