missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2A/MLL Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-256R
This item is not returnable.
View return policy
Description
KMT2A/MLL Polyclonal specifically detects KMT2A/MLL in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| KMT2A/MLL | |
| Polyclonal | |
| Western Blot, Flow Cytometry | |
| ALL1, ALL-1MLL/GAS7, CXXC7TET1-MLL, CXXC-type zinc finger protein 7, EC 2.1.1.43, HRXFLJ11783, HTRX, HTRX1MLL-AF4 der(11) fusion protein, KMT2ACDK6/MLL fusion protein, Lysine N-methyltransferase 2A, MLL/GAS7 fusion protein, MLL/GMPS fusion protein, MLL1, MLL1A, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog), myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila), Trithorax-like protein, TRX1histone-lysine N-methyltransferase MLL, Zinc finger protein HRX | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB600-256 is specific for human MLL-1 protein. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MLL | |
| The immunogen recognized by this antibody maps to a region between residues 2725 and 2775 of human myeloid/lymphoid or mixed-lineage leukemia 1 using the number given in Swiss-Prot entry Q03164 (GeneID 4297). | |
| 100 μL | |
| Transcription Factors and Regulators | |
| 4297 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction