missing translation for 'onlineSavingsMsg'
Learn More

KMT2A/MLL Antibody (BL-175-7E8), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18417135 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417135 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417135 Supplier Novus Biologicals Supplier No. NBP276397AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KMT2A/MLL Monoclonal antibody specifically detects KMT2A/MLL in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2A/MLL
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone BL-175-7E8
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias ALL1, ALL-1MLL/GAS7, CXXC7TET1-MLL, CXXC-type zinc finger protein 7, EC 2.1.1.43, HRXFLJ11783, HTRX, HTRX1MLL-AF4 der(11) fusion protein, KMT2ACDK6/MLL fusion protein, Lysine N-methyltransferase 2A, MLL/GAS7 fusion protein, MLL/GMPS fusion protein, MLL1, MLL1A, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog), myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila), Trithorax-like protein, TRX1histone-lysine N-methyltransferase MLL, Zinc finger protein HRX
Host Species Rabbit
Immunogen Immunogen sequence between amino acids 720 and 780
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4297
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.