missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KMO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100019
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KMO.
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.
Specifications
| KMO | |
| Polyclonal | |
| Unconjugated | |
| KMO | |
| AI046660; dJ317G22.1; HAMAP-Rule:MF_03018}; KMO; kmo {ECO:0000312; kynurenine 3-hydroxylase; kynurenine 3-hydroxylase {ECO:0000255; Kynurenine 3-monooxygenase; kynurenine 3-monooxygenase (kynurenine 3-hydroxylase); kynurenine 3-monooxygenase {ECO:0000255; RGD:620610} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 59113, 8564, 98256 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15229, O88867, Q91WN4 | |
| KMO | |
| A synthesized peptide derived from human KMO(Accession O15229), corresponding to amino acid residues K333-E383. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction