missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5143872
This item is not returnable.
View return policy
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Specifications
| KLRG1 | |
| Polyclonal | |
| Unconjugated | |
| Klrg1 | |
| 2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 500 μg/mL | |
| PBS with 4MG trehalose and no preservative | |
| Q96E93 | |
| Klrg1 | |
| E.coli-derived human KLRG1 recombinant protein (Position: E11-F195). | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction