missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLRG1 Monoclonal Antibody (2F1), Brilliant Violet™ 605, eBioscience™, Invitrogen™

Syrian Hamster Monoclonal Antibody
£153.00 - £321.00
Specifications
| Antigen | KLRG1 |
|---|---|
| Clone | 2F1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245103
|
Invitrogen™
406-5893-80 |
25 μg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30245419
![]() |
Invitrogen™
406-5893-82 |
100 μg |
£321.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow CytometrySpecifications
| KLRG1 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 2F1 | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| Syrian Hamster | |
| Mouse | |
| O88713 | |
| 50928 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Klrg1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title