missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLRF1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547219
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp44 and rhNKp30 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
KLRF1, an activating homodimeric C-type lectin-like receptor (CTLR), is expressed on nearly all natural killer (NK) cells and stimulates their cytoxicity and cytokine release (Kuttruff et al., 2009 ).
Specifications
| KLRF1 | |
| Polyclonal | |
| Unconjugated | |
| KLRF1 | |
| Activating coreceptor NKp80; CLEC5C; CLEC5CMGC119908; C-type lectin domain family 5 member C; killer cell lectin like receptor F1; killer cell lectin-like receptor F1; killer cell lectin-like receptor subfamily F member 1; killer cell lectin-like receptor subfamily F, member 1; KLRF1; Lectin-like receptor F1; MGC119907; MGC119909; ML; natural killer cell receptor; NKp80 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 51348 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9NZS2 | |
| KLRF1 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 C-Terminus Val66-Tyr231. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction