missing translation for 'onlineSavingsMsg'
Learn More

Klre-1 Antibody [FITC], Novus Biologicals™

Product Code. 30732986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732986 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732986 Supplier Novus Biologicals Supplier No. AF7099F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Klre-1 Polyclonal antibody specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Klre-1
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CD94
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Klre-1, Lys94-Lys226, Accession # NP_705818
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.