missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Klre-1 Polyclonal antibody specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
Specifications
| Antigen | Klre-1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD94 |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Klre-1, Lys94-Lys226, Accession # NP_705818 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?