Learn More
KLRB1 Mouse anti-Rat, APC, Clone: 10/78, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MR6805
Description
This antibody reacts with rat NKR-PIA, which is expressed by NK cells and NKT cells. Every lot is tested by flow cytometry using freshly harvested rat splenocytes. Use dim light during handling, incubation with cells and prior to analysis. It is recommended that cells be analyzed within 18 hours of staining. If the reagent is being diluted, it is recommended that only the quantity to be used within one week be diluted.
Natural Killer (NK) cells are large, granular lymphocytes found primarily in peripheral blood, where they make up about 10% of the lymphocyte population. Their function is to defend against certain types of tumor cells and virally infected cells. When triggered, the cells release cytotoxic granules which induce either apoptosis or necrosis in the target cell. There are known to be two sets of cell surface receptor on NK cells which recognize target cells with opposing functions. One set, of which an example is NKR-P1 receptors, trigger cytokine release and cell cytotoxicity and the other set, of which Ly49A is an example, inhibit cytokine release. These receptors bind with MHC class I molecules on the surface of healthy cells which inhibits the release of cytokines. In virally infected or cancerous cells, class I MHC molecules are not expressed, inducing the release of cytokine. An activation signal is known to exist, however the mechanics of this signal are not very well understood.
Specifications
| KLRB1 | |
| Monoclonal | |
| 0.2 μg/mL | |
| PBS with BSA and 0.1% sodium azide | |
| P27471 | |
| Klrb1a | |
| Rat NKR-P1A. | |
| 500 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 10/78 | |
| APC | |
| Klrb1a | |
| CD161, CLEC5B, MGC138614, NKR, NKR-P1, NKR-P1A, NKRP1A, hNKR-P1A | |
| Mouse | |
| Purified | |
| RUO | |
| 362443 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.