missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLRA3 Polyclonal Antibody

Description
A suggested positive control is mouse brain tissue lysate. PA5-20676 can be used with blocking peptide PEP-0794.
KLRA3 (also known as Ly49C) is a member of the LY49 family of receptors in Natural Killer (NK) cells that bind to major histocompatibility complex (MHC) class 1. These proteins are classified as either activating or inhibitory receptors based on whether they possess an immunoreceptor tyrosine-based inhibitory motif (ITIM) in their cytoplasmic region (for inhibitory receptors), or an immunoreceptor tyrosine-based activation motif (ITAM) that transmits activating signals resulting in phosphorylation of several substrates. KLRA3 is thought to be an inhibitory receptor, recognizing peptide -receptive H-2Kb.
Specifications
Specifications
| Antigen | KLRA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Klra3 |
| Gene Accession No. | Q64329 |
| Gene Alias | 5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor, subfamily A, member 3; Klra3; Ly49C; Ly-49c; Ly49I; Ly49I receptor; LY49I1; lymphocyte antigen 49c; natural killer cell-associated antigen 2; Nk2; Nk-2; Nk2.1; NK-2.1; T-cell surface glycoprotein Ly-49C |
| Gene Symbols | Klra3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?